Assay Design



Gene Transcripts

Gene Symbol
NFE2L1
Entrez Gene ID
403692
Gene Name
nuclear factor, erythroid 2 like 1
Gene Aliases
LCR-F1
Chromosome Location
Chr.9: 24344697 - 24356918 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005624612.3 XP_005624669.1 - 1364 97
XM_014116620.2 XP_013972095.1 - 1251 97
XM_022423058.1 XP_022278766.1 - 1250 97
XM_846772.5 XP_851865.1 - 1367 97

View Full Product Details