Assay Design



Gene Transcripts

Gene Symbol
RRP36
Entrez Gene ID
100687900
Gene Name
ribosomal RNA processing 36
Gene Aliases
-
Chromosome Location
Chr.12: 11549179 - 11553925 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003431728.4 XP_003431776.1 6 - 7 762 107

View Full Product Details