Assay Design



Gene Transcripts

Gene Symbol
NDUFA13
Entrez Gene ID
476659
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
Gene Aliases
-
Chromosome Location
Chr.20: 43721568 - 43730607 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005632651.3 XP_005632708.1 1 - 2 643 69

View Full Product Details