Assay Design
Gene Transcripts
- Gene Symbol
- IQCF1
- Entrez Gene ID
-
476604
- Gene Name
- IQ motif containing F1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 37797253 - 37799110 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_533808.6 |
XP_533808.1 |
2 - 3 |
249 |
70 |
View Full Product Details