Assay Design



Gene Transcripts

Gene Symbol
PRSS50
Entrez Gene ID
476646
Gene Name
serine protease 50
Gene Aliases
-
Chromosome Location
Chr.20: 42046177 - 42051517 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005632625.2 XP_005632682.1 4 - 5 1091 106

View Full Product Details