Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005635002.3 | XP_005635059.1 | 2 - 3 | 1383 | 84 |
XM_022409009.1 | XP_022264717.1 | 3 - 4 | 1044 | 84 | |
XM_022409010.1 | XP_022264718.1 | 3 - 4 | 1559 | 84 | |
XM_022409011.1 | XP_022264719.1 | 3 - 4 | 1558 | 84 | |
XM_022409012.1 | XP_022264720.1 | 3 - 4 | 1664 | 84 | |
XM_534409.6 | XP_534409.1 | 2 - 3 | 933 | 84 |