Assay Design



Gene Transcripts

Gene Symbol
HPF1
Entrez Gene ID
477355
Gene Name
histone PARylation factor 1
Gene Aliases
C25H4orf27
Chromosome Location
Chr.25: 20622781 - 20645459 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005635632.2 XP_005635689.1 - 491 97
XM_014107596.2 XP_013963071.1 - 554 97
XM_014107597.2 XP_013963072.1 - 542 97
XM_022409784.1 XP_022265492.1 - 510 97
XM_534550.6 XP_534550.2 - 478 97

View Full Product Details