Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005635099.2 | XP_005635156.1 | 10 - 11 | 1080 | 71 |
XM_005635103.3 | XP_005635160.1 | 10 - 11 | 1168 | 71 | |
XM_014106890.2 | XP_013962365.1 | 10 - 11 | 1067 | 71 | |
XM_022409460.1 | XP_022265168.1 | 10 - 11 | 1012 | 71 | |
XM_022409461.1 | XP_022265169.1 | 10 - 11 | 1012 | 71 | |
XM_022409462.1 | XP_022265170.1 | 9 - 10 | 940 | 71 |