Assay Design
Gene Transcripts
- Gene Symbol
- MFAP3L
- Entrez Gene ID
-
477356
- Gene Name
- microfibril associated protein 3 like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.25: 20844840 - 20863317 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_534551.6 |
XP_534551.3 |
- |
680 |
64 |
View Full Product Details