Assay Design



Gene Transcripts

Gene Symbol
PI3
Entrez Gene ID
477241
Gene Name
peptidase inhibitor 3
Gene Aliases
SKALP, trappin-2
Chromosome Location
Chr.24: 32504697 - 32506214 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290099.1 NP_001277028.1 1 - 2 93 85

View Full Product Details