Assay Design
Gene Transcripts
- Gene Symbol
- HEBP1
- Entrez Gene ID
-
477690
- Gene Name
- heme binding protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.27: 33373682 - 33398356 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_534884.5 |
XP_534884.1 |
1 - 2 |
158 |
66 |
View Full Product Details