Assay Design
Gene Transcripts
- Gene Symbol
- CLEC12A
- Entrez Gene ID
-
477697
- Gene Name
- C-type lectin domain family 12 member A
- Gene Aliases
-
-
- Chromosome Location
-
Chr.27: 36040574 - 36051249 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_534891.6 |
XP_534891.2 |
3 - 4 |
575 |
99 |
View Full Product Details