Assay Design
Gene Transcripts
- Gene Symbol
- IFI6
- Entrez Gene ID
-
478170
- Gene Name
- interferon alpha inducible protein 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 72638657 - 72642648 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_535344.6 |
XP_535344.1 |
4 - 5 |
487 |
126 |
View Full Product Details