Assay Design
Gene Transcripts
- Gene Symbol
- NDUFA2
- Entrez Gene ID
-
478032
- Gene Name
- NADH:ubiquinone oxidoreductase subunit A2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 35803452 - 35805530 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_535211.6 |
XP_535211.1 |
- |
157 |
80 |
View Full Product Details