Assay Design



Gene Transcripts

Gene Symbol
SRSF1
Entrez Gene ID
609693
Gene Name
serine and arginine rich splicing factor 1
Gene Aliases
SFRS1
Chromosome Location
Chr.9: 32692517 - 32696325 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014116687.2 XP_013972162.1 2 - 3 515 62

View Full Product Details