Assay Design



Gene Transcripts

Gene Symbol
RFX8
Entrez Gene ID
100683612
Gene Name
regulatory factor X8
Gene Aliases
-
Chromosome Location
Chr.10: 41464882 - 41521157 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423975.1 XP_022279683.1 4 - 5 341 99

View Full Product Details