Assay Design



Gene Transcripts

Gene Symbol
TREM2
Entrez Gene ID
474898
Gene Name
triggering receptor expressed on myeloid cells 2
Gene Aliases
TREML1
Chromosome Location
Chr.12: 9915692 - 9928322 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426027.1 XP_022281735.1 6 - 7 1373 62
XM_022426028.1 XP_022281736.1 6 - 7 1307 62
XM_532133.6 XP_532133.2 3 - 4 622 62

View Full Product Details