Assay Design



Gene Transcripts

Gene Symbol
TMEM14A
Entrez Gene ID
474937
Gene Name
transmembrane protein 14A
Gene Aliases
-
Chromosome Location
Chr.12: 20303724 - 20318336 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003431744.4 XP_003431792.1 2 - 3 215 102
XM_532172.6 XP_532172.3 3 - 4 243 102

View Full Product Details