Assay Design
Gene Transcripts
- Gene Symbol
- BET1
- Entrez Gene ID
-
475231
- Gene Name
- Bet1 golgi vesicular membrane trafficking protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 19490556 - 19499960 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_532463.6 |
XP_532463.1 |
3 - 4 |
368 |
140 |
View Full Product Details