Assay Design



Gene Transcripts

Gene Symbol
ATAD3A
Entrez Gene ID
479568
Gene Name
ATPase family AAA domain containing 3A
Gene Aliases
ATAD3B
Chromosome Location
Chr.5: 56587385 - 56613475 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022418903.1 XP_022274611.1 - 1462 97
XM_536708.6 XP_536708.2 - 1617 97

View Full Product Details