Assay Design



Gene Transcripts

Gene Symbol
FUS
Entrez Gene ID
100856381
Gene Name
FUS RNA binding protein
Gene Aliases
-
Chromosome Location
Chr.6: 17059904 - 17071227 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005621238.3 XP_005621295.1 - 129 56

View Full Product Details