Assay Design
Gene Transcripts
- Gene Symbol
- TIMM22
- Entrez Gene ID
-
480639
- Gene Name
- translocase of inner mitochondrial membrane 22
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 44823368 - 44830375 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_537758.6 |
XP_537758.1 |
1 - 2 |
575 |
80 |
View Full Product Details