Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003434894.4 | XP_003434942.1 | 1 - 2 | 170 | 61 |
XM_005622749.3 | XP_005622806.1 | 1 - 2 | 171 | 61 | |
XM_005622750.3 | XP_005622807.1 | 1 - 2 | 174 | 61 | |
XM_005622751.3 | XP_005622808.1 | 1 - 2 | 170 | 61 | |
XM_005622752.3 | XP_005622809.1 | 1 - 2 | 173 | 61 | |
XM_537256.6 | XP_537256.2 | 1 - 2 | 168 | 61 |