Assay Design



Gene Transcripts

Gene Symbol
SWI5
Entrez Gene ID
102152833
Gene Name
SWI5 homologous recombination repair protein
Gene Aliases
-
Chromosome Location
Chr.9: 55229510 - 55234645 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423942.1 XP_022279650.1 2 - 3 274 79

View Full Product Details