Assay Design
Gene Transcripts
- Gene Symbol
- PTRH1
- Entrez Gene ID
-
480713
- Gene Name
- peptidyl-tRNA hydrolase 1 homolog
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 55676605 - 55678442 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_537833.6 |
XP_537833.2 |
4 - 5 |
489 |
89 |
View Full Product Details