Assay Design



Gene Transcripts

Gene Symbol
ANXA8L1
Entrez Gene ID
479270
Gene Name
annexin A8-like 1
Gene Aliases
ANXA8
Chromosome Location
Chr.4: 34862992 - 34878535 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022417626.1 XP_022273334.1 6 - 7 642 75
XM_022417627.1 XP_022273335.1 6 - 7 631 75
XM_536412.6 XP_536412.2 5 - 6 539 75

View Full Product Details