Assay Design



Gene Transcripts

Gene Symbol
GABARAPL2
Entrez Gene ID
479646
Gene Name
GABA type A receptor associated protein like 2
Gene Aliases
-
Chromosome Location
Chr.5: 75230214 - 75240724 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620674.3 XP_005620731.1 3 - 4 362 107

View Full Product Details