Assay Design



Gene Transcripts

Gene Symbol
SNRPD1
Entrez Gene ID
480178
Gene Name
small nuclear ribonucleoprotein D1 polypeptide
Gene Aliases
-
Chromosome Location
Chr.7: 66407313 - 66421193 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287157.1 NP_001274086.1 2 - 3 206 152
GenBank mRNA CO672980.1 - 2 - 3 390 152
DT536786.1 - 2 - 3 192 152

View Full Product Details