Assay Design



Gene Transcripts

Gene Symbol
KCNE3
Entrez Gene ID
485197
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
-
Chromosome Location
Chr.21: 23860727 - 23872194 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014122818.2 XP_013978293.1 2 - 3 317 80
XM_014122820.2 XP_013978295.1 3 - 4 398 80
XM_014122821.2 XP_013978296.1 3 - 4 433 80
XM_014122823.2 XP_013978298.1 4 - 5 702 80
XM_022407690.1 XP_022263398.1 1 - 2 712 80
XM_022407691.1 XP_022263399.1 4 - 5 725 80

View Full Product Details