Assay Design



Gene Transcripts

Gene Symbol
ENPP4
Entrez Gene ID
481823
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases
-
Chromosome Location
Chr.12: 14353355 - 14366506 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014118485.2 XP_013973960.1 2 - 3 1016 124
XM_014118486.2 XP_013973961.1 2 - 3 1020 124

View Full Product Details