Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005631673.3 | XP_005631730.1 | 4 - 5 | 307 | 62 |
XM_005631674.3 | XP_005631731.1 | 3 - 4 | 247 | 62 | |
XM_005631676.3 | XP_005631733.1 | 2 - 3 | 362 | 62 | |
XM_005631677.3 | XP_005631734.1 | 2 - 3 | 223 | 62 | |
XM_014121268.2 | XP_013976743.1 | 3 - 4 | 414 | 62 | |
XM_014121270.2 | XP_013976745.1 | 3 - 4 | 315 | 62 | |
XM_022405822.1 | XP_022261530.1 | 4 - 5 | 341 | 62 | |
XM_022405823.1 | XP_022261531.1 | 3 - 4 | 181 | 62 | |
XM_022405824.1 | XP_022261532.1 | 4 - 5 | 394 | 62 | |
XM_022405825.1 | XP_022261533.1 | 4 - 5 | 352 | 62 | |
XM_022405826.1 | XP_022261534.1 | 4 - 5 | 360 | 62 | |
XM_022405827.1 | XP_022261535.1 | 3 - 4 | 289 | 62 | |
XM_022405828.1 | XP_022261536.1 | 3 - 4 | 255 | 62 | |
XM_540920.6 | XP_540920.2 | 4 - 5 | 519 | 62 |