Assay Design
Gene Transcripts
- Gene Symbol
- ZDHHC24
- Entrez Gene ID
-
483708
- Gene Name
- zinc finger DHHC-type containing 24
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 50801179 - 50808480 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_540829.6 |
XP_540829.2 |
1 - 2 |
435 |
87 |
View Full Product Details