Assay Design
Gene Transcripts
- Gene Symbol
- COA7
- Entrez Gene ID
-
482496
- Gene Name
- cytochrome c oxidase assembly factor 7 (putative)
- Gene Aliases
-
SELRC1
- Chromosome Location
-
Chr.15: 8706047 - 8718184 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_539613.6 |
XP_539613.2 |
2 - 3 |
312 |
117 |
View Full Product Details