Assay Design



Gene Transcripts

Gene Symbol
HRC
Entrez Gene ID
484386
Gene Name
histidine rich calcium binding protein
Gene Aliases
-
Chromosome Location
Chr.1: 107304718 - 107308741 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005616328.3 XP_005616385.1 4 - 5 2422 59

View Full Product Details