Assay Design
Gene Transcripts
- Gene Symbol
- LAMTOR1
- Entrez Gene ID
-
485211
- Gene Name
- late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.21: 25981170 - 25986942 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_542329.6 |
XP_542329.2 |
3 - 4 |
402 |
101 |
View Full Product Details