Assay Design
Gene Transcripts
- Gene Symbol
- CCDC97
- Entrez Gene ID
-
484489
- Gene Name
- coiled-coil domain containing 97
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 112648610 - 112659740 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_541603.6 |
XP_541603.3 |
1 - 2 |
200 |
108 |
View Full Product Details