Assay Design



Gene Transcripts

Gene Symbol
VGLL2
Entrez Gene ID
484097
Gene Name
vestigial like family member 2
Gene Aliases
-
Chromosome Location
Chr.1: 57529901 - 57537893 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005615589.3 XP_005615646.1 2 - 3 618 99
XM_014113304.2 XP_013968779.1 2 - 3 618 99
XM_541214.5 XP_541214.3 2 - 3 617 99

View Full Product Details