Assay Design



Gene Transcripts

Gene Symbol
MAFF
Entrez Gene ID
481255
Gene Name
MAF bZIP transcription factor F
Gene Aliases
-
Chromosome Location
Chr.10: 26475478 - 26486109 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022424449.1 XP_022280157.1 1 - 2 202 100

View Full Product Details