Assay Design



Gene Transcripts

Gene Symbol
ABT1
Entrez Gene ID
100856389
Gene Name
activator of basal transcription 1
Gene Aliases
-
Chromosome Location
Chr.35: 24357502 - 24360731 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003640168.4 XP_003640216.1 1 - 2 299 71

View Full Product Details