Assay Design



Gene Transcripts

Gene Symbol
KCNK1
Entrez Gene ID
488965
Gene Name
potassium two pore domain channel subfamily K member 1
Gene Aliases
-
Chromosome Location
Chr.4: 6117406 - 6139390 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014112737.2 XP_013968212.1 - 540 140

View Full Product Details