Assay Design



Gene Transcripts

Gene Symbol
TAF5L
Entrez Gene ID
488982
Gene Name
TATA-box binding protein associated factor 5 like
Gene Aliases
-
Chromosome Location
Chr.4: 9646516 - 9674318 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022417426.1 XP_022273134.1 - 1176 62
XM_546100.3 XP_546100.2 - 997 62

View Full Product Details