Assay Design
Gene Transcripts
- Gene Symbol
- NCEH1
- Entrez Gene ID
-
488171
- Gene Name
- neutral cholesterol ester hydrolase 1
- Gene Aliases
-
AADACL1
- Chromosome Location
-
Chr.34: 36813691 - 36874642 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_545295.6 |
XP_545295.3 |
3 - 4 |
521 |
71 |
View Full Product Details