Assay Design



Gene Transcripts

Gene Symbol
EPHX4
Entrez Gene ID
490160
Gene Name
epoxide hydrolase 4
Gene Aliases
ABHD7
Chromosome Location
Chr.6: 56863779 - 56900312 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014114843.2 XP_013970318.1 2 - 3 105 89

View Full Product Details