Assay Design
Gene Transcripts
- Gene Symbol
- SLC25A37
- Entrez Gene ID
-
486115
- Gene Name
- solute carrier family 25 member 37
- Gene Aliases
-
-
- Chromosome Location
-
Chr.25: 33923108 - 33965577 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_844354.5 |
XP_849447.1 |
1 - 2 |
407 |
75 |
View Full Product Details