Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005620995.3 | XP_005621052.1 | - | 744 | 69 |
XM_014114180.2 | XP_013969655.1 | - | 744 | 69 | |
XM_014114182.2 | XP_013969657.2 | - | 841 | 69 | |
XM_022419592.1 | XP_022275300.1 | - | 841 | 69 | |
XM_022419593.1 | XP_022275301.1 | - | 841 | 69 | |
XM_022419595.1 | XP_022275303.1 | - | 841 | 69 | |
XM_022419597.1 | XP_022275305.1 | - | 744 | 69 |