Assay Design



Gene Transcripts

Gene Symbol
SYNJ2BP
Entrez Gene ID
490757
Gene Name
synaptojanin 2 binding protein
Gene Aliases
-
Chromosome Location
Chr.8: 44044526 - 44079002 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252343.1 NP_001239272.1 3 - 4 332 62

View Full Product Details