Assay Design
Gene Transcripts
- Gene Symbol
- MMP28
- Entrez Gene ID
-
491136
- Gene Name
- matrix metallopeptidase 28
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 37873364 - 37907724 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_548256.6 |
XP_548256.2 |
1 - 2 |
366 |
68 |
View Full Product Details