Assay Design
Gene Transcripts
- Gene Symbol
- ISG20L2
- Entrez Gene ID
-
490409
- Gene Name
- interferon stimulated exonuclease gene 20 like 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 41258140 - 41263635 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_547530.5 |
XP_547530.2 |
2 - 3 |
984 |
81 |
View Full Product Details