Assay Design



Gene Transcripts

Gene Symbol
AURKAIP1
Entrez Gene ID
607067
Gene Name
aurora kinase A interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 56539987 - 56541578 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620384.3 XP_005620441.1 - 303 139
XM_014113810.2 XP_013969285.1 - 296 139
XM_843641.5 XP_848734.1 - 289 139

View Full Product Details