Assay Design
Gene Transcripts
- Gene Symbol
- CLIC3
- Entrez Gene ID
-
607042
- Gene Name
- chloride intracellular channel 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 48634061 - 48637083 on Build CanFam3.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_843599.5 |
XP_848692.1 |
1 - 2 |
1196 |
57 |
View Full Product Details